Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 12 | 1255 | ||
NM_001199114.1 | NP_001186043.1 | 11 | 1094 | |||
NM_001199115.1 | NP_001186044.1 | 11 | 1194 | |||
NM_001199116.1 | NP_001186045.1 | 11 | 1154 | |||
NM_022880.3 | NP_075018.1 | 10 | 1195 | |||
GenBank mRNA | AA407561.1 | - | 5 | 361 | ||
AF131212.1 | - | 10 | 1196 | |||
AF305501.1 | - | 11 | 1165 | |||
AK004756.1 | - | 11 | 1140 | |||
AK050089.1 | - | 11 | 1179 | |||
AK146265.1 | - | 11 | 1095 | |||
AK149483.1 | - | 11 | 1110 | |||
AK170463.1 | - | 11 | 1126 | |||
AK198320.1 | - | 2 | 141 | |||
AK212529.1 | - | 2 | 141 | |||
BC004828.1 | - | 12 | 1218 | 2631728 | ||
BC006812.1 | - | 10 | 1126 | 3155188 | ||
EU180577.1 | - | 10 | 1070 |