Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022322.2 | NP_071717.1 | 5 | 735 | ||
GenBank mRNA | AB055422.1 | - | 5 | 542 | ||
AF191768.1 | - | 5 | 681 | |||
AF219993.1 | - | 5 | 608 | |||
AF291655.1 | - | 5 | 760 | |||
AK003748.1 | - | 5 | 619 | |||
AK014761.1 | - | 5 | 574 | |||
BC006919.1 | - | 5 | 544 | 3601223 | ||
BC049944.1 | - | 5 | 735 | 2811628 |