Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083315.2 | NP_001076784.1 | 11 | 1448 | ||
NM_001289624.1 | NP_001276553.1 | 12 | 1441 | |||
NM_001289625.1 | NP_001276554.1 | 12 | 1441 | |||
NM_001289626.1 | NP_001276555.1 | 11 | 1372 | |||
NM_001289627.1 | NP_001276556.1 | 12 | 1175 | |||
NM_001289629.1 | NP_001276558.1 | 11 | 1106 | |||
NM_022332.3 | NP_071727.2 | 12 | 1517 | |||
GenBank mRNA | AF234884.1 | - | 12 | 1468 | ||
AF234885.1 | - | 11 | 1399 | |||
AK079107.1 | - | 13 | 1654 | |||
AK132103.1 | - | 11 | 1402 | |||
AK151550.1 | - | 11 | 1403 | |||
AK182928.1 | - | 2 | 105 | |||
AY007792.1 | - | 12 | 1403 | |||
AY007793.1 | - | 11 | 1334 | |||
AY007794.1 | - | 13 | 1432 | |||
AY007795.1 | - | 12 | 1363 | |||
AY007796.1 | - | 13 | 1242 | |||
AY007797.1 | - | 12 | 1173 | |||
AY007798.1 | - | 12 | 1175 | |||
AY007799.1 | - | 11 | 1106 | |||
AY551821.1 | - | 12 | 1168 | |||
AY551822.1 | - | 11 | 1099 | |||
BC012719.1 | - | 6 | 588 | |||
BC024652.1 | - | 12 | 1441 | 3660309 | ||
BC051429.1 | - | 11 | 3127 | |||
BC060630.1 | - | 12 | 3326 | |||
BC108418.1 | - | 12 | 1461 | 30603484 | ||
CT010245.1 | - | 11 | 1025 |