Gene Symbol: Stk32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022416.2 | NP_071861.1 | 9 | 1183 | ||
GenBank mRNA | AK082468.1 | - | 9 | 1184 | ||
BC052404.1 | - | 9 | 1093 | 5715729 | ||
BC056396.1 | - | 9 | 1169 | 6841272 | ||
BC058412.1 | - | 9 | 1093 | 5715729 |