Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 6 | 1053 | ||
NM_001164208.1 | NP_001157680.1 | 6 | 701 | |||
NM_001164209.2 | NP_001157681.1 | 7 | 909 | |||
NM_001286651.1 | NP_001273580.1 | 7 | 906 | |||
NM_001286652.1 | NP_001273581.1 | 6 | 813 | |||
NM_023056.4 | NP_075543.1 | 6 | 1302 | |||
GenBank mRNA | AB031386.1 | - | 6 | 862 | ||
AF115426.1 | - | 6 | 774 | |||
AK007408.1 | - | 6 | 708 | |||
AK156042.1 | - | 6 | 1054 | |||
BC019539.1 | - | 5 | 671 | 4237639 | ||
BC022166.1 | - | 6 | 1317 | 3582123 | ||
BE457679.1 | - | 2 | 366 | |||
CT010176.1 | - | 5 | 666 | |||
DV047496.1 | - | 6 | 740 | |||
DV059241.1 | - | 6 | 777 |