Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159602.1 | NP_001153074.1 | 4 | 441 | ||
NM_025376.3 | NP_079652.1 | 4 | 593 | |||
GenBank mRNA | AK003298.1 | - | 4 | 595 | ||
AK004477.1 | - | 4 | 561 | |||
BC062145.1 | - | 4 | 519 | 30286808 | ||
CX564956.1 | - | 4 | 442 |