Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003408.1 | - | 5 | 459 | ||
AK011241.1 | - | 6 | 612 | |||
AK146727.1 | - | 6 | 1087 | |||
AK151554.1 | - | 6 | 618 | |||
BC027121.1 | - | 6 | 585 | 5359960 | ||
BC033605.1 | - | 6 | 834 | 5369405 | ||
CX222514.1 | - | 6 | 644 |