Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025594.3 | NP_079870.1 | 2, 3 | 122 | ||
GenBank mRNA | AK008717.1 | - | 2, 3 | 115 | ||
AK012091.1 | - | 2, 3 | 118 | |||
BC024881.1 | - | 2, 3 | 122 | 4022326 | ||
BC089501.1 | - | 2, 3 | 117 | 6594761 |