Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025666.2 | NP_079942.1 | 9 | 1111 | ||
GenBank mRNA | AK088208.1 | - | 9 | 1113 | ||
AK146308.1 | - | 9 | 1113 | |||
AK149989.1 | - | 9 | 1066 | |||
AK155611.1 | - | 9 | 982 | |||
BC051678.1 | - | 6 | 639 | |||
BC058535.1 | - | 8 | 795 | |||
BC079531.1 | - | 8 | 808 | |||
BC094323.1 | - | 9 | 1118 | |||
BC094423.1 | - | 9 | 1069 | 6812389 |