Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025666.2 | NP_079942.1 | 6 | 597 | ||
GenBank mRNA | AK011968.1 | - | 6 | 468 | ||
AK017542.1 | - | 6 | 553 | |||
AK088208.1 | - | 6 | 599 | |||
AK149989.1 | - | 6 | 552 | |||
AK155611.1 | - | 6 | 468 | |||
AK190790.1 | - | 5 | 331 | |||
BC051678.1 | - | 3 | 125 | |||
BC058535.1 | - | 5 | 281 | |||
BC079531.1 | - | 5 | 294 | |||
BC094323.1 | - | 6 | 604 | |||
BC094423.1 | - | 6 | 555 | 6812389 |