Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023243.5 | NP_075732.1 | 4 | 565 | ||
GenBank mRNA | AF287135.1 | - | 4 | 618 | ||
AK007910.1 | - | 1 | 30 | |||
AK078059.1 | - | 4 | 566 | |||
AK137849.1 | - | 4 | 594 | |||
AK185780.1 | - | 3 | 377 | |||
AK215242.1 | - | 3 | 342 | |||
AK217691.1 | - | 4 | 245 | |||
BC038861.1 | - | 4 | 544 | 1446473 |