Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025705.3 | NP_079981.2 | 5, 6 | 685 | ||
GenBank mRNA | AK014521.1 | - | 5, 6 | 497 | ||
AK016485.1 | - | 5, 6 | 685 |