Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146243.2 | NP_666355.1 | 2 | 206 | ||
GenBank mRNA | AK032832.1 | - | 2 | 124 | ||
AK033014.1 | - | 2 | 124 | |||
AK088216.1 | - | 2 | 118 | |||
AK163217.1 | - | 2 | 119 | |||
BC027799.1 | - | 2 | 190 | 5325176 | ||
BC047530.1 | - | 2 | 105 | |||
BY278998.1 | - | 2 | 205 |