Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025755.3 | NP_080031.1 | 2, 3 | 176 | ||
GenBank mRNA | AK014276.1 | - | 2, 3 | 190 | ||
AK016966.1 | - | 2, 3 | 225 | |||
AK018101.1 | - | 2, 3 | 187 | |||
AK032498.1 | - | 1, 2 | 1924 | |||
BC018481.1 | - | 2, 3 | 176 | 3482169 |