Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025993.2 | NP_080269.1 | 3 | 892 | ||
GenBank mRNA | AK010995.1 | - | 3 | 893 | ||
AK132010.1 | - | 3 | 894 | |||
AK139957.1 | - | 4 | 611 | |||
AK154531.1 | - | 3 | 893 | |||
BC026790.1 | - | 3 | 578 | 5148857 |