Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026009.2 | NP_080285.2 | 7 | 1043 | ||
GenBank mRNA | AB040489.1 | - | 7 | 1020 | ||
AK011882.1 | - | 7 | 980 | |||
AK078812.1 | - | 7 | 1022 | |||
AK132273.1 | - | 7 | 1001 | |||
BC120502.1 | - | 7 | 931 | 40129425 | ||
BC137681.1 | - | 7 | 931 | 8860700 |