Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025695.4 | NP_079971.2 | 22 | 3084 | ||
GenBank mRNA | AJ310552.1 | - | 21 | 2676 | ||
AK003872.1 | - | 1 | 37 | |||
AK014442.1 | - | 3 | 288 | |||
AK220488.1 | - | 21 | 2759 | |||
BC026429.1 | - | 4 | 524 | |||
BC090630.1 | - | 22 | 3084 | 30536320 |