Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 10 | 1117 | ||
GenBank mRNA | AK013554.1 | - | 10 | 1069 | ||
AK021316.1 | - | 3 | 224 | |||
AK028937.1 | - | 10 | 1043 | |||
AK028945.1 | - | 10 | 1020 | |||
AK030280.1 | - | 10 | 1057 | |||
AK076409.1 | - | 1 | 51 | |||
AK134887.1 | - | 9 | 1776 | |||
AK210387.1 | - | 1 | 71 | |||
AY029531.1 | - | 10 | 1018 | |||
BC003946.1 | - | 10 | 1004 | 3487343 |