Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026157.2 | NP_080433.1 | 3 | 541 | ||
GenBank mRNA | AK002680.1 | - | 3 | 512 | ||
AK011546.1 | - | 3 | 542 | |||
AK036760.1 | - | 3 | 542 | |||
AK045203.1 | - | 3 | 531 | |||
AK076565.1 | - | 3 | 531 | |||
AK135537.1 | - | 3 | 509 | |||
AK154747.1 | - | 3 | 538 | |||
AK166038.1 | - | 3 | 519 | |||
BC017535.1 | - | 3 | 530 | |||
BC057643.1 | - | 3 | 515 | 2616306 |