Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026163.2 | NP_080439.1 | 10 | 1928 | ||
GenBank mRNA | AK004650.1 | - | 10 | 1929 | ||
AK005020.1 | - | 10 | 1960 | |||
AK049481.1 | - | 10 | 1929 | |||
AK167660.1 | - | 10 | 1930 | |||
BC018487.1 | - | 4 | 484 | |||
BC037133.1 | - | 5 | 534 | |||
BC068279.1 | - | 8 | 1208 | |||
BC138252.1 | - | 10 | 1928 | 8861271 | ||
BC138253.1 | - | 10 | 1928 | 8861272 |