Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026298.5 | NP_080574.5 | 25 | 2791 | ||
GenBank mRNA | AK043642.1 | - | 25 | 2739 | ||
AK122461.1 | - | 10 | 1085 | |||
AY339616.1 | - | 25 | 2727 | |||
BC042583.1 | - | 25 | 2730 | |||
BC066096.1 | - | 25 | 2784 | 6856438 |