Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190911.1 | NP_001177840.1 | 7 | 1161 | ||
NM_001190912.1 | NP_001177841.1 | 6 | 1083 | |||
NM_001190913.1 | NP_001177842.1 | 7 | 1161 | |||
NM_001190914.1 | NP_001177843.1 | 8 | 1262 | |||
NM_026324.3 | NP_080600.1 | 7 | 1161 | |||
GenBank mRNA | AK034142.1 | - | 7 | 1098 | ||
AK038385.1 | - | 7 | 912 | |||
AK045373.1 | - | 7 | 1066 | |||
AK079332.1 | - | 8 | 1200 | |||
AK171866.1 | - | 4 | 512 | |||
AY169782.1 | - | 7 | 1161 | |||
BC063072.1 | - | 6 | 878 | 5695003 |