Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190911.1 | NP_001177840.1 | 17 | 2397 | ||
NM_001190912.1 | NP_001177841.1 | 16 | 2394 | |||
NM_001190914.1 | NP_001177843.1 | 17 | 2462 | |||
NM_026324.3 | NP_080600.1 | 16 | 2361 | |||
GenBank mRNA | AK045373.1 | - | 17 | 2302 | ||
AK079332.1 | - | 17 | 2400 | |||
AK171866.1 | - | 13 | 1712 | |||
AY169782.1 | - | 16 | 2361 | |||
BC063072.1 | - | 16 | 2189 | 5695003 |