- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026331.3 | NP_080607.2 | 3 | 630 | ||
GenBank mRNA | AF288621.1 | - | 3 | 583 | ||
AF361699.1 | - | 2 | 2340 | |||
AK006155.1 | - | 3 | 645 | |||
AK015770.1 | - | 3 | 330 | |||
AK028588.1 | - | 1 | 1697 | |||
AK046008.1 | - | 3 | 1006 | |||
AK050414.1 | - | 3 | 598 | |||
AK162615.1 | - | 1 | 492 | |||
BC040399.1 | - | 1 | 453 | 4037680 |