Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145957.1 | NP_001139429.1 | 10 | 1324 | ||
NM_172767.3 | NP_766355.3 | 11 | 1400 | |||
GenBank mRNA | AK019914.1 | - | 11 | 1354 | ||
AK041397.1 | - | 11 | 1367 | |||
AK053665.1 | - | 11 | 1344 | |||
AK054497.1 | - | 11 | 1373 | |||
AK136934.1 | - | 11 | 1373 | |||
AK161193.1 | - | 10 | 1311 | |||
AK193979.1 | - | 2 | 171 | |||
AY366502.1 | - | 11 | 1370 | |||
BC004727.1 | - | 5 | 488 | 3582797 | ||
BC065153.1 | - | 10 | 1254 | 6418714 |