Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026539.2 | NP_080815.1 | 2, 3 | 247 | ||
GenBank mRNA | AK011061.1 | - | 2, 3 | 261 | ||
AK014473.1 | - | 2, 3 | 249 | |||
AK165656.1 | - | 2, 3 | 271 | |||
BC019617.2 | - | 2, 3 | 246 | |||
BC052385.1 | - | 2, 3 | 231 | 5703859 | ||
BC057567.1 | - | 2, 3 | 260 | 6415898 | ||
BC062966.1 | - | 2, 3 | 260 | 6415898 |