Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026620.3 | NP_080896.2 | 3 | 340 | ||
GenBank mRNA | AK019190.1 | - | 3 | 341 | ||
AK019253.1 | - | 3 | 341 | |||
AK039128.1 | - | 1481 | ||||
AK039305.1 | - | 3 | 1870 | |||
BC048087.1 | - | 3 | 330 | 6439798 |