Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212445.2 | NP_997610.1 | 5 | 1142 | ||
GenBank mRNA | AK075811.1 | - | 5 | 951 |