Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 3 | 637 | ||
NM_194051.3 | NP_918940.1 | 5 | 2924 | |||
NM_194052.3 | NP_918941.1 | 3 | 1113 | |||
NM_194053.3 | NP_918942.1 | 4 | 1170 | |||
NM_194054.3 | NP_918943.1 | 5 | 3531 | |||
GenBank mRNA | AB073672.1 | - | 3 | 1489 | ||
AF326337.1 | - | 3 | 417 | |||
AK034902.1 | - | 3 | 1057 | |||
AK211699.1 | - | 3 | 353 | |||
AK212390.1 | - | 3 | 340 | |||
AK218123.1 | - | 3 | 342 | |||
AK220511.1 | - | 5 | 3491 | |||
AY102280.1 | - | 5 | 2924 | |||
AY102281.1 | - | 3 | 1070 | |||
AY102282.1 | - | 4 | 1127 | |||
AY102283.1 | - | 3 | 630 | |||
AY102284.1 | - | 5 | 3488 | |||
AY114152.1 | - | 5 | 3489 | |||
AY114153.1 | - | 4 | 944 | |||
BC032192.1 | - | 3 | 1783 | 5323152 | ||
BC032272.1 | - | 3 | 2637 | |||
BC056373.1 | - | 5 | 3359 | |||
BU848611.1 | - | 3 | 476 |