Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 2 | 491 | ||
NM_194051.3 | NP_918940.1 | 4 | 2778 | |||
NM_194052.3 | NP_918941.1 | 2 | 967 | |||
NM_194053.3 | NP_918942.1 | 3 | 1024 | |||
NM_194054.3 | NP_918943.1 | 4 | 3385 | |||
GenBank mRNA | AB073672.1 | - | 2 | 1343 | ||
AF326337.1 | - | 2 | 271 | |||
AK034902.1 | - | 2 | 911 | |||
AK211699.1 | - | 2 | 207 | |||
AK212390.1 | - | 2 | 194 | |||
AK220511.1 | - | 4 | 3345 | |||
AY102280.1 | - | 4 | 2778 | |||
AY102281.1 | - | 2 | 924 | |||
AY102282.1 | - | 3 | 981 | |||
AY102283.1 | - | 2 | 484 | |||
AY102284.1 | - | 4 | 3342 | |||
AY114152.1 | - | 4 | 3343 | |||
AY114153.1 | - | 3 | 798 | |||
BC032192.1 | - | 2 | 1637 | 5323152 | ||
BC032272.1 | - | 2 | 2491 | |||
BC056373.1 | - | 4 | 3213 | |||
BU848611.1 | - | 2 | 330 |