Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 9 | 739 | ||
NM_152808.3 | NP_690021.1 | 9 | 741 | |||
GenBank mRNA | AK041533.1 | - | 9 | 698 | ||
AK043924.1 | - | 8 | 808 | |||
AK048648.1 | - | 9 | 726 | |||
AK088553.1 | - | 8 | 785 | |||
AK153992.1 | - | 9 | 720 | |||
AK166695.1 | - | 9 | 742 | |||
AK219076.1 | - | 2 | 122 | |||
BC013213.1 | - | 2 | 134 | |||
BC031535.1 | - | 9 | 717 | 3991609 |