Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199465.2 | NP_955759.2 | 12 | 1834 | ||
GenBank mRNA | AK135919.1 | - | 3 | 522 | ||
AK162232.1 | - | 7 | 1276 | |||
BC052878.1 | - | 12 | 1781 | 30046316 | ||
HQ179982.1 | - | 12 | 1868 | |||
HQ179983.1 | - | 10 | 1637 | |||
HQ179984.1 | - | 9 | 1466 |