Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133700.2 | NP_598461.1 | 4 | 419 | ||
GenBank mRNA | BC050781.1 | - | 4 | 419 | 6591397 |