Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133700.2 | NP_598461.1 | 11 | 1672 | ||
GenBank mRNA | BC002151.1 | - | 2 | 292 | ||
BC038876.1 | - | 2 | 238 | |||
BC039798.1 | - | 1 | 588 | |||
BC050781.1 | - | 11 | 1672 | 6591397 |