Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026873.2 | NP_081149.1 | 10 | 1071 | ||
GenBank mRNA | AK004502.1 | - | 10 | 1080 | ||
AK144396.1 | - | 10 | 1082 | |||
BC016563.1 | - | 10 | 1073 | |||
BC025110.1 | - | 10 | 1071 | 3987809 |