- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Chmp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026879.2 | NP_081155.1 | 3 | 417 | ||
GenBank mRNA | AK084111.1 | - | 3 | 419 | ||
BC042626.1 | - | 2 | 173 | |||
BC055809.1 | - | 3 | 446 | 6414077 |