Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026908.3 | NP_081184.3 | 11 | 1250 | ||
GenBank mRNA | AK013161.1 | - | 9 | 1047 | ||
AK013205.1 | - | 10 | 1243 | |||
AK030474.1 | - | 11 | 1264 | |||
AK053642.1 | - | 11 | 1251 | |||
AK076758.1 | - | 11 | 1251 | |||
AK076867.1 | - | 11 | 1271 | |||
BC016128.1 | - | 9 | 1217 | 4911640 | ||
BC016546.1 | - | 11 | 1234 | 3595339 |