Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026908.3 | NP_081184.3 | 6 | 610 | ||
GenBank mRNA | AK005323.1 | - | 609 | |||
AK013161.1 | - | 5 | 469 | |||
AK013205.1 | - | 5 | 605 | |||
AK030474.1 | - | 6 | 624 | |||
AK053642.1 | - | 6 | 611 | |||
AK076758.1 | - | 6 | 611 | |||
AK076867.1 | - | 6 | 631 | |||
BC016128.1 | - | 4 | 577 | 4911640 | ||
BC016546.1 | - | 6 | 594 | 3595339 |