Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 6 | 451 | ||
NM_001287211.1 | NP_001274140.1 | 6 | 451 | |||
NM_026972.5 | NP_081248.4 | 7 | 541 | |||
GenBank mRNA | AF373409.1 | - | 7 | 519 | ||
AF374471.1 | - | 6 | 408 | |||
AF422108.1 | - | 7 | 514 | |||
AF424790.1 | - | 7 | 513 | |||
AF424792.1 | - | 7 | 513 | |||
AF424793.1 | - | 7 | 513 | |||
AF424794.1 | - | 7 | 533 | |||
AF424795.1 | - | 7 | 530 | |||
AF424796.1 | - | 7 | 524 | |||
AF424797.1 | - | 6 | 423 | |||
AF424798.1 | - | 6 | 443 | |||
AF424799.1 | - | 6 | 423 | |||
AF424803.1 | - | 7 | 549 | |||
AK007656.1 | - | 6 | 457 | |||
AK138999.1 | - | 7 | 515 |