Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178617.4 | NP_848732.3 | 6 | 675 | ||
GenBank mRNA | AK039240.1 | - | 6 | 491 | ||
AK050307.1 | - | 7 | 572 | |||
AK081951.1 | - | 6 | 567 | |||
AY278200.2 | - | 6 | 658 | |||
BC067055.1 | - | 6 | 675 | 30545678 |