Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 1700029J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033148.3 | NP_001028320.1 | 2 | 1908 | ||
GenBank mRNA | AK156041.1 | - | 2 | 1910 | ||
BC107256.2 | - | 3 | 391 | 40052445 |