Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027135.2 | NP_081411.2 | 13 | 1722 | ||
GenBank mRNA | BC030852.1 | - | 6 | 616 | ||
BC038038.1 | - | 2 | 212 | |||
BC049257.1 | - | 2 | 115 | 5150771 | ||
BC057981.1 | - | 11 | 1515 | |||
BC067020.1 | - | 13 | 1722 | 6847085 |