Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145954.1 | NP_666066.1 | 3 | 307 | ||
GenBank mRNA | AK030673.1 | - | 3 | 300 | ||
AK144967.1 | - | 306 | ||||
AK144995.1 | - | 3 | 314 | |||
AK151691.1 | - | 3 | 307 | |||
AK161198.1 | - | 3 | 345 | |||
AK164374.1 | - | 3 | 353 | |||
AK169346.1 | - | 659 | ||||
AK170276.1 | - | 3 | 346 | |||
AK220200.1 | - | 3 | 283 | |||
BC013548.1 | - | 3 | 307 | 4222058 |