Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159349.1 | NP_001152821.1 | 1, 2 | 218 | ||
NM_027226.4 | NP_081502.2 | 1, 2 | 218 | |||
GenBank mRNA | AJ344096.1 | - | 1, 2 | 102 | ||
AK008130.1 | - | 1, 2 | 190 | |||
AK014447.1 | - | 1, 2 | 189 | |||
AK029785.1 | - | 1, 2 | 163 | |||
AK050448.1 | - | 1, 2 | 177 | |||
AK050479.1 | - | 1, 2 | 190 | |||
AK139638.1 | - | 1, 2 | 214 | |||
AK157602.1 | - | 1, 2 | 190 | |||
AK161252.1 | - | 1, 2 | 196 | |||
AK165633.1 | - | 1, 2 | 172 | |||
AK166032.1 | - | 1, 2 | 190 | |||
AK170511.1 | - | 1, 2 | 190 | |||
BC010204.1 | - | 1, 2 | 192 | 3158224 | ||
BC028253.1 | - | 1, 2 | 175 | 3983533 | ||
BY060615.1 | - | 1, 2 | 218 |