Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159349.1 | NP_001152821.1 | 3 | 436 | ||
NM_027226.4 | NP_081502.2 | 3 | 436 | |||
GenBank mRNA | AJ344096.1 | - | 3 | 320 | ||
AK008130.1 | - | 3 | 408 | |||
AK014447.1 | - | 3 | 407 | |||
AK029785.1 | - | 3 | 381 | |||
AK050448.1 | - | 3 | 395 | |||
AK050479.1 | - | 3 | 408 | |||
AK077184.1 | - | 4 | 344 | |||
AK139638.1 | - | 3 | 432 | |||
AK157602.1 | - | 3 | 408 | |||
AK161252.1 | - | 3 | 414 | |||
AK165633.1 | - | 3 | 390 | |||
AK166032.1 | - | 3 | 408 | |||
AK170511.1 | - | 3 | 408 | |||
BC010204.1 | - | 3 | 410 | 3158224 | ||
BC028253.1 | - | 3 | 393 | 3983533 |