Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027275.3 | NP_081551.2 | 18 | 1453 | ||
GenBank mRNA | AK013131.1 | - | 4 | 232 | ||
AK146005.1 | - | 18 | 1442 | |||
AK146172.1 | - | 18 | 1454 | |||
AK172480.1 | - | 19 | 1518 | |||
BC026759.1 | - | 2 | 108 | |||
BC055844.1 | - | 4 | 280 | |||
BC060687.1 | - | 9 | 674 | |||
BC084595.1 | - | 9 | 703 | |||
BC115434.2 | - | 18 | 1449 | 40094203 |