Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133786.3 | NP_598547.1 | 24 | 3969 | ||
GenBank mRNA | AJ534940.2 | - | 23 | 3845 | ||
AK049487.1 | - | 1 | 1252 | |||
BC005507.1 | - | 3 | 539 | |||
BC025542.1 | - | 4 | 641 | |||
BC061481.1 | - | 24 | 3944 | 6512832 | ||
BC062939.1 | - | 24 | 3969 | 6841276 |