Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abhd17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133722.2 | NP_598483.2 | 1, 2 | 622 | ||
GenBank mRNA | BC018511.1 | - | 1, 2 | 622 | 3483379 | |
BM943880.1 | - | 1, 2 | 103 |