Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 13 | 2016 | ||
GenBank mRNA | AK145135.1 | - | 13 | 1902 | ||
AK172920.1 | - | 13 | 1883 | |||
BC071239.1 | - | 6 | 644 | |||
BC141056.1 | - | 13 | 1976 | 9055668 | ||
CB528069.1 | - | 3 | 246 |