Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 11 | 1809 | ||
GenBank mRNA | AK139553.1 | - | 11 | 1685 | ||
AK160143.1 | - | 2 | 472 | |||
BC006674.1 | - | 11 | 1792 | 3499487 | ||
DV663229.1 | - | 1 | 15 |